siRNA Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024145.2 | NP_001019316.1 | 3, 4 | 440 | ||
| GenBank mRNA | AB195278.1 | - | 3, 4 | 440 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g4f| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024145.2 | NP_001019316.1 | 3, 4 | 440 | ||
| GenBank mRNA | AB195278.1 | - | 3, 4 | 440 | ||