siRNA Details
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013722.3 | NP_038750.2 | 10 | 1279 | ||
GenBank mRNA | AF192754.1 | - | 10 | 1224 | ||
AF498252.1 | - | 9 | 1026 | |||
AF498253.1 | - | 9 | 1026 | |||
AK080557.1 | - | 10 | 1241 | |||
BC107397.2 | - | 9 | 1145 | 40058225 |
