siRNA Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190717.1 | NP_001177646.1 | 12 | 1487 | ||
| NM_013726.3 | NP_038754.1 | 12 | 1490 | |||
| GenBank mRNA | AJ003132.1 | - | 12 | 1380 | ||
| AK132403.1 | - | 12 | 1452 | |||
| AK161782.1 | - | 1 | 471 | |||
| BC138709.1 | - | 12 | 1308 | 8861731 | ||
| BC138710.1 | - | 12 | 1308 | 8861732 | ||
| BC145392.1 | - | 12 | 1305 | 9053950 | ||

Dbf4
Genomic Map