siRNA Details
| Gene Symbol: Dok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013739.2 | NP_038767.1 | 5 | 1203 | ||
| GenBank mRNA | AF179242.1 | - | 5 | 1203 | ||
| AF237580.1 | - | 5 | 1199 | |||
| AK137520.1 | - | 5 | 1192 | |||
| AK154750.1 | - | 4 | 2026 | |||
| AK155180.1 | - | 4 | 2034 | |||
| AK156060.1 | - | 4 | 2033 | |||
| AK172511.1 | - | 4 | 2033 | |||
| BC048784.1 | - | 4 | 2334 | |||
| BC116922.1 | - | 5 | 1202 | 40126241 | ||
| BC120535.1 | - | 5 | 1202 | 40129458 | ||

Dok3
Genomic Map