siRNA Details
| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013750.2 | NP_038778.1 | 1 | 559 | ||
| GenBank mRNA | AK003767.1 | - | 1 | 525 | ||
| BC023376.1 | - | 1 | 461 | |||
| BC023408.1 | - | 1 | 461 | 5252055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phlda3| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013750.2 | NP_038778.1 | 1 | 559 | ||
| GenBank mRNA | AK003767.1 | - | 1 | 525 | ||
| BC023376.1 | - | 1 | 461 | |||
| BC023408.1 | - | 1 | 461 | 5252055 | ||