siRNA Details
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013753.2 | NP_038781.2 | 5 | 828 | ||
| GenBank mRNA | AK088577.1 | - | 5 | 664 | ||
| AK146489.1 | - | 5 | 773 | |||
| BC052850.1 | - | 674 | ||||
| BC080827.1 | - | 4 | 619 | |||
| BC099531.1 | - | 5 | 643 | |||
| BC137695.1 | - | 4 | 592 | 8860714 | ||
| BC144846.1 | - | 4 | 598 | 9053394 | ||
| BC144847.1 | - | 4 | 598 | 9053395 | ||
| X99384.1 | - | 4 | 613 | |||

Pald1
Genomic Map