siRNA Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013759.2 | NP_038787.1 | 2 | 147 | ||
| GenBank mRNA | AF195142.1 | - | 2 | 122 | ||
| AK002339.1 | - | 2 | 107 | |||
| AK002652.1 | - | 2 | 106 | |||
| AK003112.1 | - | 2 | 107 | |||
| AK003624.1 | - | 2 | 107 | |||
| BC090646.1 | - | 2 | 147 | 30946285 | ||
| BC141146.1 | - | 2 | 111 | 9055758 | ||
| CX566836.1 | - | 2 | 147 | |||

Msrb1
Genomic Map