siRNA Details
| Gene Symbol: Srr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163311.1 | NP_001156783.1 | 5 | 631 | ||
| NM_013761.4 | NP_038789.1 | 6 | 857 | |||
| GenBank mRNA | AF148321.1 | - | 5 | 512 | ||
| AK017235.1 | - | 6 | 736 | |||
| AK028034.1 | - | 5 | 514 | |||
| AK031687.1 | - | 6 | 729 | |||
| AK043738.1 | - | 6 | 763 | |||
| AK079343.1 | - | 2 | 141 | |||
| AK157122.1 | - | 5 | 782 | |||
| AK170096.1 | - | 6 | 743 | |||
| AK170471.1 | - | 6 | 743 | |||
| BC011164.1 | - | 6 | 734 | 4195695 | ||

Srr
Genomic Map