siRNA Details
| Gene Symbol: Mmel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013783.2 | NP_038811.2 | 8 | 1024 | ||
| GenBank mRNA | AF157105.1 | - | 8 | 970 | ||
| AF157106.1 | - | 6 | 679 | |||
| AF176569.1 | - | 8 | 1021 | |||
| AF302075.1 | - | 6 | 645 | |||
| AF302076.1 | - | 7 | 714 | |||
| AF302077.1 | - | 6 | 645 | |||
| AK154366.1 | - | 5 | 513 | |||
| BC137649.1 | - | 6 | 797 | 8860668 | ||
| BC144804.1 | - | 6 | 797 | 9053352 | ||

Mmel1
Genomic Map