siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013785.2 | NP_038813.2 | 6 | 1457 | ||
| GenBank mRNA | AF177144.1 | - | 5 | 1157 | ||
| AK037925.1 | - | 5 | 1267 | |||
| AK088872.1 | - | 6 | 1475 | |||
| AK171310.1 | - | 6 | 1875 | |||
| AK172921.1 | - | 6 | 1516 | |||
| BC053030.1 | - | 6 | 1461 | |||
| BC059006.1 | - | 6 | 1457 | 6827203 | ||

Ip6k1
Genomic Map