siRNA Details
| Gene Symbol: Pdhx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175094.5 | NP_780303.1 | 11 | 1415 | ||
| GenBank mRNA | AK047670.1 | - | 11 | 1413 | ||
| AK050433.1 | - | 11 | 1416 | |||
| AK083170.1 | - | 11 | 1379 | |||
| BC061231.1 | - | 11 | 1376 | 6532760 | ||

Pdhx
Genomic Map