siRNA Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013852.2 | NP_038880.1 | 7 | 775 | ||
| GenBank mRNA | AK030626.1 | - | 7 | 740 | ||
| BC032923.1 | - | 7 | 716 | 1430389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcf3| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013852.2 | NP_038880.1 | 7 | 775 | ||
| GenBank mRNA | AK030626.1 | - | 7 | 740 | ||
| BC032923.1 | - | 7 | 716 | 1430389 | ||