siRNA Details
| Gene Symbol: Abca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039581.2 | NP_001034670.1 | 23 | 4120 | ||
| NM_013855.3 | NP_038883.2 | 24 | 4193 | |||
| GenBank mRNA | AK089794.1 | - | 24 | 4137 | ||
| AK154698.1 | - | 24 | 4101 | |||
| AK171239.1 | - | 6 | 1234 | |||
| AY083616.1 | - | 24 | 4088 | |||
| BC042663.1 | - | 18 | 3135 | |||
| BC079617.1 | - | 18 | 2919 | 30362309 | ||
| CV784401.1 | - | 4 | 672 | |||

Abca3
Genomic Map