siRNA Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110273.1 | NP_001103743.1 | 11 | 1996 | ||
| NM_207651.3 | NP_997534.2 | 22 | 3136 | |||
| GenBank mRNA | AK032837.1 | - | 13 | 1740 | ||
| BC150680.1 | - | 22 | 3044 | 9087591 | ||

Slc14a2
Genomic Map