siRNA Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018795.2 | NP_061265.2 | 24 | 3340 | ||
| GenBank mRNA | BC040400.1 | - | 24 | 3348 | ||
| BC049980.1 | - | 8 | 1283 | |||
| EF109740.1 | - | 24 | 3340 | |||
| EF109741.1 | - | 24 | 3340 | |||

Abcc6
Genomic Map