siRNA Details
| Gene Symbol: Shroom3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077595.2 | NP_001071063.1 | 8 | 5032 | ||
| NM_001077596.2 | NP_001071064.1 | 8 | 5080 | |||
| NM_015756.2 | NP_056571.2 | 9 | 5404 | |||
| GenBank mRNA | AF199421.1 | - | 9 | 5422 | ||
| AF199422.1 | - | 8 | 4990 | |||
| AK049347.1 | - | 5 | 2938 | |||
| AK129371.1 | - | 5 | 4410 | |||
| AK147493.1 | - | 8 | 5015 | |||
| AK164918.1 | - | 8 | 4991 | |||
| BC003909.1 | - | 3 | 676 | |||
| BC145113.1 | - | 10 | 4821 | 9053661 | ||

Shroom3
Genomic Map