siRNA Details
| Gene Symbol: 1700088E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138581.2 | NP_613047.1 | 2, 3 | 215 | ||
| GenBank mRNA | AA008475.1 | - | 2, 3 | 204 | ||
| AK006539.1 | - | 2, 3 | 196 | |||
| BC068203.1 | - | 1, 2 | 260 | |||
| BC096672.1 | - | 1, 2 | 313 | 5117646 | ||
| BC116891.1 | - | 2, 3 | 146 | 40126210 | ||
| BC116893.1 | - | 2, 3 | 146 | 40126212 | ||
| BC144714.1 | - | 2, 3 | 146 | 9053262 | ||

1700088E04Rik
Genomic Map