siRNA Details
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033568.2 | NP_291046.1 | 6 | 615 | ||
| GenBank mRNA | AK013058.1 | - | 6 | 617 | ||
| AK020203.1 | - | 4 | 305 | |||
| BC003938.1 | - | 6 | 587 | 3486703 | ||
| BC030490.1 | - | 1 | 580 | |||

Snf8
Genomic Map