siRNA Details
| Gene Symbol: Trp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025246.1 | NP_001020417.1 | 8 | 766 | ||
| GenBank mRNA | AK142316.1 | - | 9 | 942 | ||
| BC024710.1 | - | 6 | 1100 | |||
| BC036165.1 | - | 3 | 249 | |||
| BC048398.1 | - | 8 | 678 | 6397829 | ||
| BC059041.1 | - | 7 | 1003 | |||
| BC098206.1 | - | 8 | 766 | 6812412 | ||

Trp53i11
Genomic Map