siRNA Details
| Gene Symbol: Slc15a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177787.4 | NP_808455.2 | 3 | 717 | ||
| GenBank mRNA | AK036413.1 | - | 2 | 500 | ||
| BC141332.1 | - | 2 | 436 | 9055944 | ||
| BC141334.1 | - | 2 | 436 | 9055946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc15a5| Gene Symbol: Slc15a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177787.4 | NP_808455.2 | 3 | 717 | ||
| GenBank mRNA | AK036413.1 | - | 2 | 500 | ||
| BC141332.1 | - | 2 | 436 | 9055944 | ||
| BC141334.1 | - | 2 | 436 | 9055946 | ||