siRNA Details
| Gene Symbol: Gm715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271548.1 | NP_001258477.1 | 1 | 359 | ||
| GenBank mRNA | CJ044530.1 | - | 1 | 360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm715| Gene Symbol: Gm715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271548.1 | NP_001258477.1 | 1 | 359 | ||
| GenBank mRNA | CJ044530.1 | - | 1 | 360 | ||