siRNA Details
| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023665.3 | NP_076154.3 | 4 | 910 | ||
| GenBank mRNA | AB041555.1 | - | 5 | 1641 | ||
| AK003799.1 | - | 3 | 2214 | |||
| AK012363.1 | - | 4 | 911 | |||
| AK028145.1 | - | 3 | 2215 | |||
| AK029529.1 | - | 3 | 2211 | |||
| AK150024.1 | - | 4 | 911 | |||
| AK150665.1 | - | 4 | 890 | |||
| AK152736.1 | - | 4 | 890 | |||
| AK160993.1 | - | 3 | 2212 | |||
| AK169598.1 | - | 5 | 1666 | |||
| AK169760.1 | - | 3 | 2213 | |||
| AK178549.1 | - | 3 | 256 | |||
| AK206224.1 | - | 4 | 301 | |||
| BC043057.1 | - | 3 | 2197 | 6419597 | ||
| BC046231.1 | - | 3 | 2115 | 6813388 | ||
| BC054728.1 | - | 3 | 2185 | 6419723 | ||
| BC056986.1 | - | 3 | 2146 | 6816649 | ||

Rsrp1
Genomic Map