siRNA Details
| Gene Symbol: Miip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025365.2 | NP_001020536.2 | 8 | 1341 | ||
| GenBank mRNA | AK133976.1 | - | 10 | 1493 | ||
| AK170224.1 | - | 7 | 1463 | |||
| BC017525.1 | - | 7 | 1413 | 4910951 | ||
| BC024069.1 | - | 8 | 1123 | 5097088 | ||
| CO039431.1 | - | 3 | 486 | |||
| FJ618908.1 | - | 8 | 1341 | |||

Miip
Genomic Map