siRNA Details
| Gene Symbol: Kiss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178260.3 | NP_839991.2 | 2 | 183 | ||
| GenBank mRNA | AB162440.1 | - | 2 | 145 | ||
| AB666166.1 | - | 3 | 343 | |||
| AF472576.1 | - | 2 | 195 | |||
| BC117046.1 | - | 2 | 180 | 40126365 | ||
| BC119348.1 | - | 2 | 180 | 8734097 | ||

Kiss1
Genomic Map