siRNA Details
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178260.3 | NP_839991.2 | 2 | 346 | ||
GenBank mRNA | AB162440.1 | - | 2 | 308 | ||
AB666166.1 | - | 3 | 506 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178260.3 | NP_839991.2 | 2 | 346 | ||
GenBank mRNA | AB162440.1 | - | 2 | 308 | ||
AB666166.1 | - | 3 | 506 |