siRNA Details
| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023746.4 | NP_076235.1 | 4 | 478 | ||
| GenBank mRNA | AF226611.1 | - | 4 | 425 | ||
| AK005447.1 | - | 4 | 479 | |||
| BC051672.1 | - | 4 | 465 | 30017843 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl5a1| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023746.4 | NP_076235.1 | 4 | 478 | ||
| GenBank mRNA | AF226611.1 | - | 4 | 425 | ||
| AK005447.1 | - | 4 | 479 | |||
| BC051672.1 | - | 4 | 465 | 30017843 | ||