siRNA Details
| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081122.1 | NP_001074591.1 | 2 | 236 | ||
| NM_001301689.1 | NP_001288618.1 | 2 | 236 | |||
| GenBank mRNA | AK076549.1 | - | 2 | 237 | ||
| KC211185.1 | - | 2 | 93 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep63| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081122.1 | NP_001074591.1 | 2 | 236 | ||
| NM_001301689.1 | NP_001288618.1 | 2 | 236 | |||
| GenBank mRNA | AK076549.1 | - | 2 | 237 | ||
| KC211185.1 | - | 2 | 93 | |||