siRNA Details
| Gene Symbol: Slco1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013797.5 | NP_038825.1 | 1 | 359 | ||
| GenBank mRNA | AB031813.1 | - | 1 | 200 | ||
| AF148218.1 | - | 1 | 183 | |||
| AF223067.1 | - | 1 | 193 | |||
| AK041125.1 | - | 1 | 207 | |||
| AK165360.1 | - | 1 | 214 | |||
| AY195868.1 | - | 1 | 192 | |||
| AY195869.1 | - | 1 | 192 | |||
| AY243579.1 | - | 1 | 192 | |||
| BB848260.1 | - | 1 | 359 | |||
| BC041147.1 | - | 1 | 142 | 4975986 | ||

Slco1a1
Genomic Map