siRNA Details
| Gene Symbol: Sbsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083903.1 | NP_001077372.1 | 1 | 121 | ||
| NM_172205.3 | NP_757342.2 | 1 | 121 | |||
| GenBank mRNA | AK081760.1 | - | 1 | 56 | ||
| AY115494.1 | - | 1 | 60 | |||
| AY444556.1 | - | 1 | 60 | |||
| BC051531.1 | - | 1 | 130 | |||
| BC092350.1 | - | 1 | 50 | |||
| BC139385.1 | - | 1 | 59 | 8862407 | ||

Sbsn
Genomic Map