siRNA Details
| Gene Symbol: Bag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013863.5 | NP_038891.4 | 4 | 1324 | ||
| GenBank mRNA | AB041583.1 | - | 4 | 1291 | ||
| AF130471.1 | - | 4 | 1057 | |||
| AK007414.1 | - | 4 | 1135 | |||
| AK016510.1 | - | 4 | 1131 | |||
| AK036675.1 | - | 4 | 1133 | |||
| AK171734.1 | - | 4 | 1266 | |||
| AK191626.1 | - | 1 | 50 | |||
| BC145765.1 | - | 4 | 1237 | 40131068 | ||

Bag3
Genomic Map