siRNA Details
| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013866.2 | NP_038894.2 | 6 | 733 | ||
| GenBank mRNA | AK137748.1 | - | 6 | 740 | ||
| BC017644.1 | - | 6 | 790 | 4504518 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp385a| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013866.2 | NP_038894.2 | 6 | 733 | ||
| GenBank mRNA | AK137748.1 | - | 6 | 740 | ||
| BC017644.1 | - | 6 | 790 | 4504518 | ||