siRNA Details
| Gene Symbol: Olfr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019473.1 | NP_062346.1 | 1 | 479 | ||
| GenBank mRNA | BC130252.1 | - | 1 | 483 | 40142475 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr155| Gene Symbol: Olfr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019473.1 | NP_062346.1 | 1 | 479 | ||
| GenBank mRNA | BC130252.1 | - | 1 | 483 | 40142475 | |