siRNA Details
| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013876.3 | NP_038904.1 | 1, 2 | 397 | ||
| GenBank mRNA | AB024427.1 | - | 2, 3 | 262 | ||
| AF220206.1 | - | 1, 2 | 151 | |||
| AK014254.1 | - | 1, 2 | 551 | |||
| AK178138.1 | - | 12 | ||||
| AK183757.1 | - | 12 | ||||
| AK210997.1 | - | 12 | ||||
| BC010299.1 | - | 1, 2 | 397 | 3155804 | ||
| BC028255.1 | - | 1, 2 | 578 | 4972336 | ||

Rnf11
Genomic Map