siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159630.1 | NP_001153102.1 | 14 | 1976 | ||
| NM_013883.2 | NP_038911.1 | 14 | 1976 | |||
| GenBank mRNA | AB030906.1 | - | 14 | 1875 | ||
| AB221680.1 | - | 10 | 1517 | |||
| AB221681.1 | - | 10 | 1517 | |||
| AK009416.1 | - | 2 | 134 | |||
| BC025478.1 | - | 5 | 2218 | |||
| BC034667.1 | - | 12 | 1686 | 5356322 | ||

Scmh1
Genomic Map