siRNA Details
| Gene Symbol: Dnajc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253685.1 | NP_001240614.1 | 4, 5 | 499 | ||
| NM_013888.3 | NP_038916.1 | 4, 5 | 636 | |||
| GenBank mRNA | AF132906.2 | - | 4, 5 | 585 | ||
| AK002649.1 | - | 2, 3 | 324 | |||
| AK032207.1 | - | 3, 4 | 2005 | |||
| BC013501.1 | - | 4, 5 | 543 | 4222828 | ||
| BC071211.1 | - | 3, 4 | 1901 | |||
| BF453706.1 | - | 4, 5 | 499 | |||
| DV042983.1 | - | 4, 5 | 651 | |||

Dnajc12
Genomic Map