siRNA Details
| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 2 | 306 | ||
| GenBank mRNA | AK017652.1 | - | 2 | 287 | ||
| BC036997.1 | - | 2 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp292| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 2 | 306 | ||
| GenBank mRNA | AK017652.1 | - | 2 | 287 | ||
| BC036997.1 | - | 2 | 318 | |||