siRNA Details
| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013889.2 | NP_038917.2 | 8 | 5540 | ||
| GenBank mRNA | AB093243.1 | - | 1 | 3112 | ||
| AB221608.1 | - | 1 | 1953 | |||
| AF017806.1 | - | 1 | 4010 | |||
| BC008537.1 | - | 1 | 1628 | |||
| BC064316.1 | - | 1 | 45 | |||

Zfp292
Genomic Map