siRNA Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153547.6 | NP_705775.2 | 10 | 1202 | ||
| GenBank mRNA | AK077523.1 | - | 10 | 1133 | ||
| AK167183.1 | - | 10 | 1086 | |||
| AY181025.1 | - | 10 | 1060 | |||
| AY185498.1 | - | 10 | 1060 | |||
| BC018560.1 | - | 10 | 1041 | |||
| BC037996.1 | - | 10 | 1062 | 5052858 | ||

Gnl3
Genomic Map