siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 5, 6 | 371 | ||
| GenBank mRNA | AK029489.1 | - | 4, 5 | 196 | ||
| AK037748.1 | - | 1, 2 | 653 | |||
| AK050534.1 | - | 5, 6 | 381 | |||
| AK086617.1 | - | 5, 6 | 405 | |||
| BC008086.1 | - | 5, 6 | 371 | 3586161 | ||
| BC030675.1 | - | 5, 6 | 369 | 1068780 | ||
| BC037025.1 | - | 1, 2 | 1728 | |||
| BC083081.1 | - | 5, 6 | 405 | 6510396 | ||

Zfp330
Genomic Map