siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 7 | 570 | ||
| GenBank mRNA | AK029489.1 | - | 6 | 395 | ||
| AK037748.1 | - | 3 | 852 | |||
| AK050534.1 | - | 7 | 580 | |||
| AK086617.1 | - | 7 | 604 | |||
| BC008086.1 | - | 7 | 570 | 3586161 | ||
| BC030675.1 | - | 7 | 568 | 1068780 | ||
| BC037025.1 | - | 3 | 1927 | |||
| BC083081.1 | - | 7 | 604 | 6510396 | ||

Zfp330
Genomic Map