siRNA Details
| Gene Symbol: Slc24a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175034.3 | NP_778199.2 | 3 | 380 | ||
| GenBank mRNA | AB085629.1 | - | 3 | 383 | ||
| AK089225.1 | - | 3 | 384 | |||
| AY236527.1 | - | 3 | 350 | |||
| BC094232.1 | - | 3 | 340 | 30538386 | ||
| BC144866.1 | - | 3 | 375 | 9053414 | ||
| BC145643.1 | - | 3 | 375 | 40130946 | ||

Slc24a5
Genomic Map