siRNA Details
| Gene Symbol: Hist1h2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175658.2 | NP_783589.1 | 1 | 118 | ||
| GenBank mRNA | BC107354.1 | - | 1 | 118 | 40055855 | |
| BC107355.1 | - | 1 | 118 | 40055860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2aa| Gene Symbol: Hist1h2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175658.2 | NP_783589.1 | 1 | 118 | ||
| GenBank mRNA | BC107354.1 | - | 1 | 118 | 40055855 | |
| BC107355.1 | - | 1 | 118 | 40055860 | ||