siRNA Details
| Gene Symbol: Sctr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012322.2 | NP_001012322.2 | 8 | 1089 | ||
| GenBank mRNA | AK032652.1 | - | 8 | 1091 | ||
| BC089355.1 | - | 7 | 990 | 30551620 | ||
More Information
Set Membership: |
|

Sctr
Genomic Map