siRNA Details
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012322.2 | NP_001012322.2 | 3 | 482 | ||
GenBank mRNA | AK032652.1 | - | 3 | 484 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
