siRNA Details
| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176902.3 | NP_795876.1 | 2 | 347 | ||
| GenBank mRNA | AK050732.1 | - | 2 | 229 | ||
| AK144901.1 | - | 2 | 349 | |||
| BC116983.1 | - | 2 | 218 | 40126302 | ||
| BC119080.1 | - | 2 | 219 | 8733829 | ||
| BY734584.1 | - | 2 | 349 | |||

Ubald2
Genomic Map