siRNA Details
| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176902.3 | NP_795876.1 | 3 | 464 | ||
| GenBank mRNA | AK050732.1 | - | 3 | 346 | ||
| AK144901.1 | - | 3 | 466 | |||
| BC116983.1 | - | 3 | 335 | 40126302 | ||
| BC119080.1 | - | 3 | 336 | 8733829 | ||
| BY734584.1 | - | 3 | 466 | |||

Ubald2
Genomic Map