siRNA Details
| Gene Symbol: C230057M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032654.1 | - | 6 | 1719 | ||
| AK082503.1 | - | 2 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230057M02Rik| Gene Symbol: C230057M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032654.1 | - | 6 | 1719 | ||
| AK082503.1 | - | 2 | 319 | |||