siRNA Details
| Gene Symbol: B230216G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242345.1 | NP_001229274.1 | 5 | 232 | ||
| GenBank mRNA | AK043509.1 | - | 3 | 145 | ||
| AK045625.1 | - | 2 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230216G23Rik| Gene Symbol: B230216G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242345.1 | NP_001229274.1 | 5 | 232 | ||
| GenBank mRNA | AK043509.1 | - | 3 | 145 | ||
| AK045625.1 | - | 2 | 326 | |||