siRNA Details
| Gene Symbol: 6430573F11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176952.4 | NP_795926.2 | 3 | 288 | ||
| GenBank mRNA | AK032504.1 | - | 5 | 1006 | ||
| AK038497.1 | - | 5 | 728 | |||
| AK039253.1 | - | 4 | 638 | |||
| AK089162.1 | - | 5 | 957 | |||
| AK141601.1 | - | 662 | ||||
| AK220392.1 | - | 1 | 141 | |||
| BC052184.1 | - | 3 | 288 | 6336362 | ||

6430573F11Rik
Genomic Map