siRNA Details
| Gene Symbol: Celf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176954.3 | NP_795928.2 | 6 | 635 | ||
| GenBank mRNA | AK029796.1 | - | 6 | 1809 | ||
| AK083461.1 | - | 5 | 363 | |||
| AK132321.1 | - | 6 | 601 | |||
| AK158669.1 | - | 5 | 412 | |||
| BC079845.1 | - | 6 | 607 | |||

Celf5
Genomic Map